Cimarec™ Basic Hotplates, Thermo Scientific
Supplier: Thermo Fisher Scientific
Certificates
About this item
A sleek, rugged design and construction reduces the chance of top plate breakage and promotes durability, for economical high performance.
- Reach maximum temperature with in 8 minutes
- Solid ceramic top plate cleans easily, resists acids and alkalis
- New top-plate design reduces chance of breakage
- Spillaway design diverts spills away from internal components
- Control knob allows precise temperature control
- Integral ring-stand holder accommodates 1.3 cm (1/2") diameter support rod
Customers Who Bought This Also Bought