Matrix™ Reagent Reservoirs, Thermo Scientific
Catalog #: CA101505-066
Supplier: Thermo Fisher Scientific
Specifications
- Capacity:25 ml
- Sterility:Sterile
- Color:White
- Description:Reagent Reservoir with Divider
- Material:Polystyrene
- Cat. no.:CA101505-066
- Packaging:10 Bags of 10
- Supplier No.:8095
Specifications
About this item
Matrix™ Reagent Reservoirs are sterile and offer an extra-wide base to add rigidity and stability, helping to avoid spills.
- Trough within a trough maximizes amount of liquid accessible to pipet tips
- Pour-off spouts on all four corners reduce spillage
- Graduations on inside wall enable quick measurement
- Sterile reservoirs are packaged in snap and tear bags, maintaining sterility while being easy to open
Customers Who Bought This Also Bought