Order Entry
ContactUsLinkComponent
DuPont™ Tyvek® 600 Coveralls with Standard Hood, Type 5-B and 6-B Chemical Protection
DuPont™ Tyvek® 600 Coveralls with Standard Hood, Type 5-B and 6-B Chemical Protection
Catalog #: CA10016-732
Supplier:  DuPont
Coverall With Hood 2Xl
DuPont™ Tyvek® 600 Coveralls with Standard Hood, Type 5-B and 6-B Chemical Protection
Catalog #: CA10016-732
Supplier:  DuPont

Specifications

  • Clothing Size:
    XXL
  • Garment application:
    Industrial,
    Chemical protection,
    Laboratory
  • Color:
    White
  • Cat. no.:
    CA10016-732
  • Chest Size:
    30"
  • Inseam:
    34"
  • Sleeve:
    393/4"

Specifications

About this item

Tyvek® 600 garments are composed of flash spun high density polyethylene which creates a unique, nonwoven material available only from DuPont.

  • Standard fit hood
  • Serged and over-taped seams
  • Elastic thumb loops
  • Elastic wrists and ankles
  • Elastic face opening
  • Glued-in elastic waist
  • Self-adhesive Tyvek® storm flap
  • Self-adhesive chin flap

Tyvek® garments provide an ideal balance of protection, durability and comfort. They are now available with serged and over-taped seams that provides the performance of Type 4/5/6 garments, which have been tested to standards against heavy liquid aerosols and airborne solid particles. The coveralls provide an effective barrier against many water-based inorganic chemicals in low concentration and particles (down to 1.0 micron in size). Tyvek® 600 coveralls are suitable for applications such as pharmaceutical manufacturing, medical applications, research and biosecurity laboratories, nuclear and maintenance. Fabric and seams offer chemical permeation barrier to low concentration water-based inorganic chemicals. Serged and over-taped seams offer extra protection and strength. Self-adhesive chin flap allows for a tight seal of suit to the mask. Elastic face, wrists and ankles as well as glued-in waist elastic allow for a tighter fit. Elastic thumb loops keep sleeves in place.

Certifications: Thickness meets DIN EN ISO 534. Chemical protective clothing, Category III, Type 4-B, 5-B and 6-B. EN 1073-2 (protection against radioactive contamination). Protection against infective agents (EN 14126) including resistance to penetration by blood and body fluids using synthetic blood (ISO 16603). Antistatic treatment (EN 1149-5) on both sides.
Customers Who Bought This Also Bought