Order Entry
ContactUsLinkComponent
NEFA-HR2 Color A
NEFA-HR2 Color A
Catalog #: CA97000-010
Supplier:  Wako Life Sciences
CAS Number:  
NEFA HR (2) COLOR REAGENT A
NEFA-HR2 Color A
Catalog #: CA97000-010
Supplier:  Wako Life Sciences
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Description:
    NEFA-HR(2) Color A
  • Size:
    4 x 50 mL
  • Method:
    Enzymatic
  • Storage temperature:
    2-10°C
  • Linearity:
    0.01 - 4.00 mEq/L
  • Stability:
    30 days after reconstitution
  • Supplier No.:
    999-34691

Specifications

About this item

The Wako HR series NEFA-HR(2) is an in vitro enzymatic colorimetric method assay for the quantitative determination of non-esterified fatty acids (NEFA) in serum.

See order tab for additional NEFA HR(2) reagents, linearity set, and calibrators.

Ordering information: Compatible with NEFA-HR2 Color B, NEFA-HR2 Solvent A, NEFA-HR2 Solvent B, Nefa Standard Solution, and NEFA Linearity Set.
Customers Who Bought This Also Bought