Thermolyne™ Industrial Benchtop Muffle Furnaces, Thermo Scientific
Supplier: Thermo Fisher Scientific
Certificates
About this item
Furnaces ideal for gravimetric analysis, heat treatment
- Rugged design with multiple safety features and choice of two temperature control options
- Reaches 1200 °C maximum temperature
- Heavy-duty firebrick insulation designed to surround the opening for added durability
- Thermocouple break protection cuts power to the heating elements and prevents thermocouple failure
- Counter-weighted door swings upward, directing heat away from operator
- LED display simultaneously shows both setpoint and actual furnace temperatures in either °C or °F
- 1 year warranty (parts and labor)
It features adjustable alarm or over-temperature protection (OTP) setting can be used to protect the furnace or loaded chamber from excessive heat and the four individual embedded elements in special refractory cement permit excellent heat distribution in the chamber.
Certifications: All units are cUL, UL listed.
Ordering information: It includes Platinel® II thermocouple and a ceramic hearth tray to protect the bottom heating element.
Customers Who Bought This Also Bought