59-801 Cut Protection Sleeves, Cane Mesh, Ansell
Supplier: Ansell Healthcare
Certificates
About this item
With double thickness (Two Ply), these flexible cane mesh sleeves provide excellent protection against chemicals, snags, punctures and abrasions.
- Excellent protection against chemicals and punctures
- Good cut resistance
- Ambidextrous
Use in the chemical processing, electronics manufacturing, food processing, food service, forestry, natural gas, oil refining, and pharmaceutical industries. Uses include exposure to chemicals, solvents, hazardous liquids, PVC cements or standing water, food packing and handling, food processing, general manufacturing, and meat packaging.
Specifications
- Catalog No:
- 80093-326
- 80093-324
- 80093-322
- Model:
- 59-801
- 59-801
- 59-801
- Garment application:
- Industrial, Laboratory
- Industrial, Laboratory
- Industrial, Laboratory
- Gender:
- Unisex
- Unisex
- Unisex
- Fabric:
- Polyester
- Polyester
- Polyester
- Thickness:
- 0.13 cm (1/16")
- 0.13 cm (1/16")
- 0.13 cm (1/16")