59-201 Liquid Protection Sleeve, Polyethylene, Ansell
Catalog #: 80093-402
Supplier: Ansell Healthcare
Specifications
- Garment application:Food,Industrial,Chemical protection,Oils, fats and fuels
- Garment fit:Regular
- Gender:Unisex
- Length:40.6 cm (16")
- Color:Blue
- Fabric:Polyethylene
- Thickness:0.8 mil
- Cat. no.:80093-402
- Type:59-201
- Supplier No.:951819
Specifications
About this item
Embossed polyethylene sleeves provide excellent protection against chemicals, snags, punctures, and abrasions.
- Excellent low-cost protection
- Provides resistance to chemicals, fats, oils, and grease
- Elasticized sleeves opening are completely sealed with no exposed elastic
- Ambidextrous
Use in the chemical processing, electronics manufacturing, food processing, food service, forestry, natural gas, oil refining, and pharmaceutical industries. Uses include exposure to chemicals, solvents, hazardous liquids, PVC cements or standing water, food packing and handling, food processing, general manufacturing, and meat packaging.
Customers Who Bought This Also Bought