Urea 98.0-102.0% USP, Ph. Eur., BP, JP endotoxin tested
Supplier: Avantor Performance Materials
Certificates
About this item
Synonyms: Carbonyl diamide,Carbamide,Diaminomethanone
Formula:NH₂CONH₂
MW:60.06 g/mol
Boiling Pt:197 °C (1013 hPa)
Melting Pt:132.7 °C
Density:1.34 g/cm³ (20 °C)
MDL Number:MFCD00008022
CAS Number:57-13-6
:Length:
:Width:
:Height:
:Ø
:Thickness:
:98,0-102,0
:Boiling Pt:
:Boiling Pt: 197 °C (1013 hPa)
:Melting Pt:
:Melting Pt: 132.7 °C
:Flash Pt:
:Flash Pt:
:M.W. 60.06 g/mol
:132.7
:197
:12,10005
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specification Test Results
- Alcohol-insoluble matter:≤ 0.04%
- Assay:98.0 – 102.0%
- Bacterial Endotoxins:≤ 2.5 EU/g
- Heavy Metals 231:≤ 0.002%
- Identification A (USP):PASS
- Identification B (USP):PASS
- Organic Impurities: Any unspecified impurity:≤ 0.1%
- Organic Impurities: Total Impurities:≤ 2.0%
- Organic Impurities: Urea related compound A:≤ 0.1%
- Residue on Ignition 281:≤ 0.1%
- Alkalinity Test:PASS
- Ammonium (2.4.1):≤ 500 ppm
- Appearance of Solution:PASS
- Assay (dried substance):98.5 – 101.5%
- Biuret:≤ 0.1%
- Heavy Metals (2.4.8):≤ 10 ppm
- Identification A: Melting Point:132° – 135°C
- Identification B: IR:PASS
- Loss on Drying (2.2.32):≤ 1.0%
- Sulfated Ash (2.4.14):≤ 0.1%
- Assay (JP):99.0%
- Chloride 1.03:≤ 0.007%
- Ethanol - Insoluble Substances:≤ 2.0 mg
- Heavy Metals 1.07:≤ 20 ppm
- Identification (1):PASS
- Identification (2):PASS
- Melting Point 2.60:132.5° – 134.5°C
- Residue on Ignition 2.44:≤ 0.1
- Sulfate 1.14:≤ 0.010%
- Urea 98.0-102.0% USP, Ph. Eur., BP, JP endotoxin tested:
Specification Test Results
Related Information
Certificates
Search by Lot NumberAT-UREA_VWRB568_PRODUCT SPECIFICATION_rev3.pdf spec_VWRB568-12KG spec_VWRB568-500G spec_VWRB568-50KG
Product specification
ghs_VWRB568
SDS
Customers Who Bought This Also Bought