Calcium chloride dihydrate 99.0-107.0% USP, Ph. Eur., BP, JP endotoxin tested
Supplier: Avantor Performance Materials
Certificates
About this item
Synonyms: Calcium Chloride Dihydrate
Formula:CaCl₂·2H₂O
MW:147.01 g/mol
Boiling Pt:1600 °C (1013 hPa)
Melting Pt:176 °C
Density:1.85 g/cm³ (20 °C)
MDL Number:MFCD00149613
CAS Number:10035-04-8
:Length:
:Width:
:Height:
:Ø
:Thickness:
:99,0-107,0
:Boiling Pt:
:Boiling Pt: 1600 °C (1013 hPa)
:Melting Pt:
:Melting Pt: 176 °C
:Flash Pt:
:Flash Pt:
:M.W. 147.01 g/mol
:Calcium chloride dihydrate
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specification Test Results
- Assay (USP):99.0 – 107.0%
- Endotoxin (USP):≤ 2.5EU/g
- Heavy Metals 231 (USP):≤ 10ppm
- Identification A (USP):PASS
- Identification B (USP):PASS
- Iron, Aluminum, and Phosphate (USP):PASS
- Limit of Magnesium and Alkali Salts (USP):≤ 1.0%
- pH (50mg/mL, Water) @ 25C (USP):4.5 – 9.2
- Acidity or Alkalinity (EP & BP):PASS
- Aluminum Test (EP & BP):PASS
- Appearance of Solution (EP & BP):PASS
- Assay (EP) (EP & BP):97.0 – 103.0%
- Barium Test (EP & BP):PASS
- Heavy Metals (2.4.8) (EP & BP):≤ 20ppm
- Identification A (EP & BP):PASS
- Identification B (EP & BP):PASS
- Identification C (EP & BP):PASS
- Iron (2.4.9) (EP & BP):≤ 10ppm
- Magnesium and Alkali Metals (EP & BP):≤ 0.5%
- Sulfates (2.4.13) (EP & BP):≤ 300ppm
- Arsenic 1.11 (JP):≤ 2ppm
- Assay (JP):96.7% - 103.3%
- Barium Test (JP):PASS
- Clarity and Color of Solution (JP):PASS
- Heavy Metals 1.07 (JP):≤ 10ppm
- Hypochlorite (JP):PASS
- Identification (JP):PASS
- Iron, Aluminum or Phosphate Test (JP):PASS
- pH 2.54 (JP):4.5 – 9.2
- Sulfate 1.14 (JP):≤ 0.024%
- Calcium chloride dihydrate 99.0-107.0% USP, Ph. Eur., BP, JP endotoxin tested:
Specification Test Results
Related Information
Certificates
Search by Lot Number Customers Who Bought This Also Bought