Thermolyne™ Benchtop Muffle Furnaces, Thermo Scientific
Supplier: Avantor
Certificates
About this item
Furnaces ideal for gravimetric analysis, ashing of organic and inorganic samples, sintering, quantitative analysis and heat treating.
- Efficient by reducing energy consumption and increase heatup time
- Reaches a 1200 °C maximum temperature
- Thermocouple break protection cuts power to heating elements and prevents thermocouple from failure
- Two open coil heating elements on chamber sides assure fast heat-up with minimum temperature gradient
- Thermal-efficient ceramic insulation surrounds chamber for maximum energy efficiency
- F47900 models have 2 L chamber capacity and F48000 models have 5 L chamber capacity
- 1 year warranty (parts and labor)
Built-in vent port removes contaminants and moisture to extend the life of the heating element and furnace. For added protection, the door safety switch stops power to heating elements when door opens.
Certifications: CSA approved and CE marked.
Ordering information: It includes power cord and one hearth tray, F48000 models also include a ceramic shelf.
Customers Who Bought This Also Bought