About this item
These nylon-reinforced aprons feature 7-millimeter polyurethane with sewn edges and offer users a combination of chemical protection, flexibility and toughness.
- Comfortable, adjustable fit
- Durable construction
- Designed for comfort and extended wear
Wherever oil, grease and chemicals are present, Ansell aprons help workers keep clean and protected from harmful contact
Three-piece 28" ties and sealed grommets provide a durable, consistent and adjustable fit on all sizes and styles. Sealed grommets allow for size adjustment without tear-out. 56-600 aprons are designed for comfort and extended wear. Use in the chemical processing, electronics manufacturing, food processing, medical laboratories, natural gas, oil refining, pharmaceutical, and research laboratory industries. Uses include food handling and preparation, industrial maintenance, and chemical processing.
Polyurethane aprons offer superior flexibility and excellent resistance to chemicals, fats, oils and grease.
Specifications
- Catalog No:
- 80093-354
- 80093-352
- Model:
- 56-600 Series
- 56-600 Series
- Garment application:
- Medical, Food, Industrial, Chemical protection, Oils, fats and fuels, Electronics/semi-conductor, Laboratory, Pharmaceutical
- Medical, Food, Industrial, Chemical protection, Oils, fats and fuels, Electronics/semi-conductor, Laboratory, Pharmaceutical
- Gender:
- Unisex
- Unisex
- Fabric:
- Urethane/Nylon
- Urethane/Nylon
- Thickness:
- 7 mil
- 7 mil