ContactUsLinkComponent
POLYTRON® Homogenizer PT 10-35 GT Package, Kinematica
POLYTRON® Homogenizer PT 10-35 GT Package, Kinematica
Supplier:  Avantor
Certificates
POLYTRON® Homogenizer PT 10-35 GT Package, Kinematica
POLYTRON® Homogenizer PT 10-35 GT Package, Kinematica
Supplier:  Avantor
Certificates
Specifications
  • Catalog No:
  • 97036-086
  • Speed range:
  • 30000 rpm max.
  • Dimensions:
  • Ø: 5-36 mm Generator
  • Power rating:
  • 1200W