Ptec™ Ionizing Air Nozzle, Transforming Technologies
Supplier: Transforming Technologies
Certificates
About this item
A specialized piezoceramic high voltage transformer drives the technology making Ptec™ ionizers among the most reliable ionizers available
Ptec™ ionizers save time and money because they require no calibration. Emitter life is increased and maintenance frequency is reduced, due to 68KHz AC ion emission.
The model IN3425 air ionizing nozzle produces a 68KHz AC output of approximately 2200V and a continuous stream of balanced air ions. The IN3425 works efficiently with clean dry air (CDA) and nitrogen (N2), and is capable of 100psi input. Stable, small, and free from EMI, the IN3425 is equipped with auto ion balance and will alarm if the high voltage output fails.
Specifications
- Catalog No:
- 10837-116
- 10837-114
- Air volume:
- 28 - 100 psi
- 28 - 100 psi
- Decay time:
- 1000 - 100 V, < 2 s at 6”
- 1000 - 100 V, < 2 s at 6”