Specifications
- Description:SRM500K Digital Surface Resistance Test Kit
- Cat. no.:10837-050
- Supplier No.:SRM500K
Specifications
About this item
The SRM500K is a digital surface resistance test kit designed to test all conductive, anti-static, and static dissipative surfaces for electrical resistivity/resistance according to EOS/ESD, CECC, ANSI, ASTM test procedures
Standard features include 10V and 100V range voltage selection, automatic zeroing, and power shut off. The unit includes two 2.3kg (5lb.) disk probes, built-in resistivity probes, and carrying case. The device offers a test range of 10³ to 10¹². Readout is provided on an LCD alpha-numeric scale, without LEDs.
It accurately and quickly measures resistance between two points (RTT) and surface to ground resistance (RTG). Suitable for factory audits and test lab evaluations, the OHM Metrics™ SRM500K also measures ambient temperature and humidity.