OHM Metrics™ Wrist Strap and Foot Testing Station, Transforming Technologies
Catalog #: 10837-038
Supplier: Transforming Technologies
Specifications
- Description:GTS600K Wrist Strap/Foot Tester and Foot Plate
- Cat. no.:10837-038
- Supplier No.:GTS600K
Specifications
About this item
The GTS600K tester is a space saving, versatile combo grounding tester that allows for accurate and cost effective testing of ESD ground devices
Dual test circuits: test footwear or wrist straps
Positive rocker switch LED's lights and audible alarm signal test results
Wrist Strap Range: 750 Kilohms – 10 Megohms
Footwear Range: 750 Kilohms – 100 Megohms
Unit does not offer simultaneous independent footwear testing
The dual test circuits in the GTS600K gives the option for both wrist strap and footwear testing. A positive rocker switch makes test mode selection easy. This tester fully supports the “Compliance Verification Plan” requirements as stated in the ANSI ESD S20.20.
Customers Who Bought This Also Bought