Cytospin™ Collection Fluid, Epredia™
Supplier: Thermo Scientific
Certificates
About this item
Store specimens or unstained slides for weeks without adverse effect utilizing this collection fluid.
- Packaged in convenient pump bottle or pre-filled specimen cups
Ready-to-use alcohol/Carbowax™, (PEG) combination for direct specimen collection and fixation.
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.