Specifications
- Description:Periodic Acid Solution (0.5%)
- Cat. no.:CA84000-254
- Supplier No.:88016
Specifications
About this item
Demonstrate the presence of both neutral and acidic mucosubstances with this stain kit.
Can be used alone to stain acid mucins and other carboxylated and sulfated acid mucosubstances blue on tissue sections. Periodic Acid Schiff (PAS) reaction is used to stain glycogen, basement membranes and neutral mucosubstances.