Double Cytoslide™ Micrsocope Slides, Epredia
Supplier: Epredia
Certificates
About this item
Use Epredia™ Double Cytoslides with Shandon™ EZ Double Cytofunnel™ Chambers and Epredia™ Cytospin™ cytocentrifuge.
- Specimen is displayed within a circle marked as 1 or 2
- Number circles correspond with the Cytofunnel™ chamber
Ordering information: Sold in boxes of 100 slides.
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.