Cylinders, Natural, Narrow Mouth, HDPE, C&G Containers
Supplier: Avantor
Certificates
About this item
Bottles are made up of high-density polyethylene (HDPE) which is used for the collection of aqueous media for testing metals and a variety of inorganic parameters, including alkalinity, anions, acidity, cyanide, sulfate, and hardness
- Container measurements and capacity 'contain' volumes may vary
A lot certificate of analysis is generated with the analytical results obtained, including the concentration reporting limits and units of measurement, for Pre-cleaned and Certified products. This certificate of analysis is included in each product case.
Secondary uses for these containers include containment and archiving.
Certifications: ASTM Type I and National Environmental Laboratory Accreditation Program Certified.
Ordering information: Pre-cleaned and Certified products contain the Certificate of Analysis and Cleaning Protocol Form in the product case.
Customers Who Bought This Also Bought