VWR® Contour™ ADA Height Base Cabinets, Acid Storage
Supplier: VWR International
Certificates
About this item
Heavy-gauge steel full-frame construction, specifically designed for the storage of corrosive chemicals.
- Smooth, easy-to-maintain surfaces
- Phosphate-coated with a baked, chemical-resistant, powder-coat finish
- Manufactured of prime grade, cold-rolled, U.S. Standard sheet steel with no ragged edges
- Lined with a molded one piece linear LDPE tub with coved corners and a 1" lip with 1⁄8" PE sheet
- Doors are latched using a nylon roller catch
Each cabinet is furnished with a PVC vent pipe for venting to the fume hood above
A 2.54 cm (1") thick work surface creates an overall height of 33". All cupboard cabinets have one full-width adjustable shelf.
(Requires a 2" hole in the hood work-surface).
Customers Who Bought This Also Bought