To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- pH:7.4
- Description:TE buffer, 1X with 0.01 M Tris, 0.001 M EDTA
- Size:500 ml
- Cat. no.:CA12001-412
- Supplier No.:51235
Specifications
About this item
AccuGENE™ Molecular Biology Buffers are ready-to-use solutions ideal for a wide range of molecular biology applications.
- Reliable - Manufactured according to strict quality control standards to ensure lot-to-lot consistency
- High Quality - No detectable DNase, RNase, or protease activities unless otherwise noted
- Efficient - Ready-made solutions eliminate experiment preparation time.
- Flexible - Customized solutions are available to meet individual needs
- Composition - 10 mM Tris, 1 mM EDTA (disodium salt), pH 7.4
TE buffer has traditionally been used to solubilize DNA or RNA, while protecting it from degradation. A common buffer, 'TE' is derived from its components: Tris, a common pH buffer, and EDTA, a molecule that chelates cations like Mg2+. Lonza’s AccugeneTM buffer is the perfect choice for DNA/RNA storage or any of the many other applications TE buffer is used for.
Store all reagents at 18 to 26 °C (64 to 79 °F).
18 megOhm water is filtered using a 0.2 µm filter, and filled into sterile bottles.