Specifications
- Length:9"
- Description:Mayo Guyon Kidney Clamp
- Material:Stainless Steel
- Working Surface Style:Serrated
- Curvature:Curved
- Cat. no.:10195-700
- Supplier No.:33-1090
Specifications
About this item
The Sklar® Mayo-Guyon Kidney Clamp is most commonly used in urological procedures where heavy tissue or organs need to be held
The heavy serrations are across the entire length of the jaws and can grasp thicker tissue.