VWR® Contour™ Wall Mounted Storage Cabinets with Open Storage
Supplier: VWR International
Certificates
About this item
Heavy-gauge steel full-frame construction.
- Smooth, easy-to-maintain surfaces
- Phosphate-coated with a baked, chemical-resistant, powder-coat finish
- Manufactured of prime grade, cold-rolled, U.S. Standard sheet steel with no ragged edges
Wall cabinets come with two adjustable shelves.
Ordering information: Installation is required.
Customers Who Bought This Also Bought