Imipenem : Cilastatin, powder
Catalog #: AAJ66272-MB
Supplier: Thermo Scientific Chemicals
CAS Number:
Specifications
- Size:25 mg
- Cat. no.:AAJ66272-MB
- Supplier No.:J66272-MB
Specifications
About this item
:Length:
:Width:
:Height:
:Ø
:Thickness:
:Boiling Pt:
:Boiling Pt:
:Melting Pt:
:Flash Pt:
:Flash Pt:
Storage Temperature:Refrigerator
Customers Who Bought This Also Bought