Tubocurarine chloride pentahydrate 98+%
Catalog #: AAJ60222-MC
Supplier: Thermo Scientific Chemicals
CAS Number: 6989-98-6
Specifications
- Size:100 mg
- Cat. no.:AAJ60222-MC
- Supplier No.:J60222-MC
Specifications
About this item
Synonyms: Curare
Melting Pt:268 °C (dec.)
ADR:"6.1",III
:Length:
:Width:
:Height:
:Ø
:Thickness:
:98+%
:98,0
:Boiling Pt:
:Boiling Pt:
:Melting Pt:
:Melting Pt: 268 °C (dec.)
:Flash Pt:
:Flash Pt:
:M.W. 681.65 g/mol
Formula:C₃₈H₄₄Cl₂N₂O₆
MW:681.65 g/mol
MDL Number:MFCD00013471
EINECS:615-023-7
:268 °C
CAS Number:6989-98-6
UN:1544
:"6.1"
:III
:GHS06
Storage Temperature:Refrigerator
:14,9807
Customers Who Bought This Also Bought