VWR® Filling and Venting Cap for VWR® Carboys
Catalog #: 10820-438
Supplier: VWR International
Specifications
- Cap Size:83 mm
- Description:Filling/Venting 3-Port Cap for VWR® Carboys
- Material:PP/silicone
- Cat. no.:10820-438
- Cap Color:Purple
Specifications
About this item
The Filling and Venting Cap for VWR® Carboys is an autoclavable closure designed for the aseptic transfer of media, biological reagents, pure water, and chemicals
Each cap is equipped with integrated 1/4" ports.
Customers Who Bought This Also Bought