About this item
Designed and developed as a breathable glove, MaxiFlex® Ultimate™ has become the benchmark for precision handling in dry environments.
- Ultra cool and comfortable to wear
- Excellent dexterity, sensitivity, and tactility
- Designed to produce minimal lint and dust
- Minimal penetration keeps inner liner soft
With applications in general assembly, micro-engineering, automotive components, painting, and horticulture, these gloves offer a wide array of uses in diverse applications. The micro foam nitrile coatings are compatible with light oils and will provide a good grip and excellent abrasion resistance. Knit wrists help prevent dirt and debris from entering the glove. The patented micro foam nitrile coating offers 360° breathability, and the 'hand at rest' design reduces hand fatigue and increases comfort. Gloves are washable in tempered water not exceeding 40 °C (104 °F).
These seamless knit nylon/lycra gloves feature a nitrile coated micro foam grip on the palm, fingers, and knuckles.
Specifications
- Catalog No:
- 20002-366
- Ambidextrous/Hand-specific:
- Ambidextrous
- Disposable / reusable:
- Reusable
- Latex/Latex-free:
- Latex-free
- Powdered/Powder-free:
- Powder-free