Specifications
- Length:6 1/2"
- Description:Mead Rongeur, 4×13 mm Bite
- Disposable / reusable:Reusable
- Latex/Latex-free:Latex-free
- Material:Stainless Steel
- Curvature:Curved
- Cat. no.:10195-640
- Supplier No.:40-3208
Specifications
About this item
Mead spring handle rongeur is used in orthopedic procedures for handling or cutting small bones. The curved jaw beaks ensures a firm grip on the bone and eases gouging areas that may have otherwise been harder to reach.