Specifications
- Description:Collin Hartmann Retractor
- Material:Stainless Steel
- Cat. no.:10197-702
- Supplier No.:60-1087
Specifications
About this item
Sklar's® Collin Hartmann Retractor is a double-ended, "S" shaped, hand held retractor commonly used in dental procedures to hold back the lips, cheeks, or tongue
It is a wireform retractor with a straight handling area and two angled ends for retracting. One end of the retractor measures 52 x 33 mm. The other measures 30 x 21 mm. The ends of the retractor are rounded and smooth.