Voltage and Electrical Advertencia Signs, National Marker
Catalog #: 94021-946
Supplier: National Marker
Specifications
- Material:Rigid plastic
- Legend:Riesgo De Electricidad No Tocar
- Dimensions:25.4×35.6 cm (10×14")
- Supplier No.:SPW500RB
- Header:Adventencia
Specifications
About this item
Safety signs are the perfect way to alert your employees and guests when a hazard is present in your facility
Customers Who Bought This Also Bought