PYREX® Separatory Squibb Funnel, with PYREX® Stopper and Rotaflo Stopcock, Corning
Supplier: Corning
Certificates
About this item
Made of PYREX® borosilicate glass.
- Very good chemical resistance
- Very good temperature resistance
- Stem Ext.Ø. is 10 mm
These separatory funnels have Rotaflo® stopcocks and strong standard taper glass stoppers.
Specifications
- Catalog No:
- 30356-449
- 30356-482
- 30356-460
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- I-RECs, ISO 14001 - Monterrey
- I-RECs, ISO 14001 - Monterrey
- I-RECs, ISO 14001 - Monterrey