To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:chemokine (C-C motif) ligand 13
- Clonality:Monoclonal
- Clone:G7_89.3
- Gene ID:Q99616
- Host:Mouse
- Neutralisation:Yes
- Reactivity:Human
- Antigen symbol:CCL13
- Conjugation:Unconjugated
- ELISA:Yes
- Size:0.5 mg
- Western blot:Yes
- Form:Lyophilized
- Antigen synonyms:CCL13
- Storage temperature:Stable at room temperature for at least one month and for greater than a year when kept at –20 °C. Reconstitute in sterile water to a concentration of 0.1 - 1.0 mg/mL. Stable for at least six weeks at 2-4 °C when reconstituted.
- Shipping temperature:RT
- Immunogen:The monoclonal antibody was produced in BALB/c mice using recombinant human MCP-4 as the immunizing antigen.
- Purification:This IgG1/K antibody was purified from a Protein A chromatography column.
- Cat. no.:10813-680
- Supplier No.:XP-5731-M
Specifications
About this item
MCP4 is a member of the b-chemokine family (C-C) of Cytokines. Like Eotaxin, MCP4 is a powerful eosinophil chemo-attractant. Additionally, like MCP-3, MCP-4 elicits chemo-attraction of monocytes and T lymphocytes
Neutralization: To yield one-half maximal inhibition [ND50] of the biological activity of rMCP-4 (100 ng/mL), a concentration of 1-5 ug/mL of this antibody is required. ELISA: In a sandwich ELISA (assuming 100 uL/well), a concentration of 2.0-4.0 ug/mL of this antibody will detect at least 100 pg/mL of recombinant human MCP-4 when used with our biotinylated antigen affinity purified anti-human MCP-4 (XP-5225Bt) as the detection antibody at a concentration of approximately 0.5-1.0 ug/mL. Western Blot: To detect hMCP-4 by Western Blot analysis this antibody can be used at a concentration of 0.20-0.40 ug/mL. Used in conjunction with compatible secondary reagents the detection limit for recombinant hMCP-4 is 2.0-4.0 ng/lane, under non-reducing conditions.
Type: Primary
Antigen: CCL13
Clonality: Monoclonal
Clone: G7_89.3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human