To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:protein kinase D1
- Clonality:Polyclonal
- Gene ID:Q15139
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Antigen symbol:PRKD1
- Conjugation:Unconjugated
- Size:0.1 mg
- Western blot:Yes
- Form:Liquid
- Antigen synonyms:PRKD1
- Modification:Tyr 463
- Storage buffer:Antibody supplied in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.
- Molecular weight:130 kDa
- Storage temperature:Store antibody at –20 °C for up to one year.
- Concentration:1 mg/mL
- Shipping temperature:4 °C
- Immunogen:PKD1/PKC μ (phospho Tyr463) antibody was raised against a peptide sequence around phosphorylation site of Tyrosine463 (R-Y-Y (p)-K-E) derived from Human PKD1/PKC μ.
- Purification:Antibodies were purified by affinity-chromatography using epitope-specific peptide.
- Cat. no.:10814-210
- Supplier No.:80-011
Specifications
About this item
Serine/threonine-protein kinase that converts transient diacylglycerol
(DAG) signals into prolonged physiological effects downstream of PKC, and is involved in
the regulation of MAPK8/JNK1 and Ras signaling, Golgi membrane integrity and
trafficking, cell survival through NF-kappa-B activation, cell migration, cell differentiation
by mediating HDAC7 nuclear export, cell proliferation via MAPK1/3 (ERK1/2) signaling,
and plays a role in cardiac hypertrophy, VEGFA-induced angiogenesis, genotoxic-induced
apoptosis and flagellin-stimulated inflammatory response. Phosphorylates the epidermal
growth factor receptor (EGFR) on dual threonine residues, which leads to the
suppression of epidermal growth factor (EGF)-induced MAPK8/JNK1 activation and
subsequent JUN phosphorylation. Phosphorylates RIN1, inducing RIN1 binding to 14-3-3
proteins YWHAB, YWHAE and YWHAZ and increased competition with RAF1 for binding
to GTP-bound form of Ras proteins (NRAS, HRAS and KRAS). Acts downstream of the
heterotrimeric G-protein beta/gamma-subunit complex to maintain the structural
integrity of the Golgi membranes, and is required for protein transport along the
secretory pathway. In the trans-Golgi network (TGN), regulates the fission of transport
vesicles that are on their way to the plasma membrane. May act by activating the lipid
kinase phosphatidylinositol 4-kinase beta (PI4KB) at the TGN for the local synthesis of
phosphorylated inositol lipids, which induces a sequential production of DAG,
phosphatidic acid (PA) and lyso-PA (LPA) that are necessary for membrane fission and
generation of specific transport carriers to the cell surface. Under oxidative stress, is
phosphorylated at Tyr-463 via SRC-ABL1 and contributes to cell survival by activating IKK
complex and subsequent nuclear translocation and activation of NFKB1. Involved in cell
migration by regulating integrin alpha-5/beta-3 recycling and promoting its recruitment
in newly forming focal adhesion.
Western Blot: 1:500~1:1000
Type: Primary
Antigen: PRKD1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat