To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:v-erb-b2 avian erythroblastic leukemia viral oncogene homolog 2
- Clonality:Polyclonal
- Gene ID:P04626
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Antigen symbol:ERBB2
- Conjugation:Unconjugated
- Size:0.1 mg
- Western blot:Yes
- Form:Liquid
- Antigen synonyms:ERBB2
- Modification:Tyr 1112
- Storage buffer:Antibody supplied in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.
- Molecular weight:150 kDa
- Storage temperature:Store antibody at –20 °C for up to one year.
- Concentration:1 mg/mL
- Shipping temperature:4 °C
- Immunogen:HER2 (phospho Tyr1112) antibody was raised against a peptide sequence around phosphorylation site of tyrosine 1112 (Q-R-Y (p)-S-E) derived from Human HER2.
- Purification:Antibodies were purified by affinity-chromatography using epitope-specific peptide.
- Cat. no.:10814-114
- Supplier No.:79-955
Specifications
About this item
Essential component of a neuregulin-receptor complex, although neuregulins do not interact with it alone. GP30 is a potential ligand for this receptor. Not activated by EGF, TGF-alpha and amphiregulin.
Western Blot: 1:500~1:1000
Type: Primary
Antigen: ERBB2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat