Precision™ Shaking Water Baths, Thermo Scientific
Supplier: THERMO FISHER SCIENTIFIC MX
Certificates
About this item
Thermo Scientific™ Precision™ shaking water baths support a range of sensitive life science and QA/QC applications – from warming fragile reagents to tissue culturing and genetics sequencing
Easily clean and maintain your bath with new coil-free interior. The Precision shallow form shaking bath has a removable tray that allows for use with smaller sample containers. Precision Dubnoff shaking water baths are designed specifically for applications that require your samples to be incubated in a controlled atmosphere.
Certifications: cULus Listed, CE compliant, US FDA Class I Medical Device.
Ordering information: Included Stainless Steel Lid, Shaking Platform, Power Cord, Yellow Duck.
Specifications
- Catalog No:
- 10805-314
- 10805-312
- 10805-310
- Model:
- 15 L
- 15 L
- 27 L
- Temperature stability:
- ±0.1°C
- ±0.1°C
- ±0.1°C
- Temperature uniformity:
- ±0.05°C
- ±0.05°C
- ±0.05°C
- Heating capacity:
- 1200 W
- 1200 W
- 1500 W
- Capacity:
- 15 L (4.0 gal.)
- 15 L (4.0 gal.)
- 27 L (7.1 gal.)
- Electrical:
- 100-115V/200-230V, 50/60Hz
- 100-115V/200-230V, 50/60Hz
- 100-115V/200-230V, 50/60Hz
- Ext. W×D×H:
- 39.4L x 63.2W x 24.9H cm (15.5 x 24.9 x 9.8")
- 39.4L x 63.2W x 24.9H cm (15.5 x 24.9 x 9.8")
- 39.4L x 93.8W x 24.9H cm (15.5 x 36.9 x 9.8")
Specifications
Customers Who Bought This Also Bought