Precision™ Circulating Water Baths, Thermo Scientific
Supplier: Thermo Fisher Scientific
Certificates
About this item
Thermo Scientific™ Precision™ circulating water baths are ideal when temperature uniformity and control are particularly critical, such as when working with enzymes and in serological applications
Available in three different models, these high performance baths feature a microprocessor control and include a removable stainless steel gable cover, diffuser tray, and rubber duck.
Certifications: cULus Listed, CE compliant, US FDA Class I Medical Device.
Ordering information: Include Stainless Steel Lid, Diffuser Tray, Power Cord, Yellow Duck.
Specifications
- Catalog No:
- 10805-386
- 10805-390
- Model:
- 19 L
- 89 L
- Capacity:
- 19 L (5.0 gal.)
- 89 L (23.5 gal.)
- Temperature stability:
- ±0.1°C
- ±0.1°C
- Temperature uniformity:
- ±0.05°C
- ±0.05°C
- Heating capacity:
- 1200 W
- 1500 W
- Electrical:
- 100-115V/200-230V, 50/60Hz
- 100-115V/200-230V, 50/60Hz
- Ext. W×D×H:
- 39.4 x 63.2 x 24.9 cm (15.5 x 24.9 x 9.8")
- 54.6L x 116W x 30H cm (21.5 x 45.7 x 11.8")
Specifications
Customers Who Bought This Also Bought