About this item
The Slide-A-Lyzer® G2 Dialysis Cassette offers efficiency, convenience and sample protection in one package. Sample loading and removal are easily accomplished by using a serological pipette or hypodermic needle attached to a syringe. The built-in air chamber provides sample buoyancy and vertical orientation of the cassette during dialysis.
- Pipette-accessible for easy sample loading and retrieval
- Self-floating chambers for buoyancy and vertical orientation during dialysis
- Designed to maintain the highest possible sample integrity and protection with rigorous quality testing for maximum consistency
- Fast and consistent dialysis with maximum sample recovery
- Ideal for removing low-molecular weight contaminants, performing buffer exchange and desalting
- Store at room temperature
The cassette membrane is composed of low-binding regenerated cellulose for maximum sample recovery while maintaining maximum sample purity and are manufactured using clean-room conditions to ensure cassettes are contaminant-free.
Volume range using a pipette: 15 to 30 ml
Volume range using a syringe: 10 to 30 ml
They are available in four precise membrane MWCOs (2K, 3.5K, 10K and 20K) for dialysing sample volumes up to 30 ml.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Related Information
Certificates
Search by Lot NumberThermo_Scientific_Complete_Protein_Clean-Up_Toolbox_Brochure