Potassium dihydrogen phosphate 98.0-100.5% (dried basis) NF, ACS, J.T.Baker®
Catalog #: CAJT7390-19
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
CAS Number: 7778-77-0
MOC: RegulatedCertificates
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Packaging:PE pail, screw top
- Size:12 kg
- Cat. no.:CAJT7390-19
Specifications
Specification Test Results
- Meets ACS Reagent Chemical Requirements:
- GMP Manufactured Product:
- Meets N.F. Requirements:
- CAUTION: For Manufacturing, processing or repackaging:
- Bulk Pharmaceutical Chemical:
- NF - Assay (KH₂PO₄) (dried basis):98.0 - 100.5 %
- NF - Arsenic (As):≤3 ppm
- NF - Fluoride (F):≤10 mg/kg
- NF - Identification A:Passes Test
- NF - Identification B:Passes Test
- NF - Insoluble Substances:≤0.2 %
- NF - Lead (Pb):≤5 ppm
- NF - Loss on Drying at 105°C:≤1.0 %
- ACS - Assay (KH₂PO₄) (by acidimetry):≥99.0 %
- ACS - Chloride (Cl):≤0.001 %
- ACS - Heavy Metals (as Pb):≤0.001 %
- ACS - Insoluble Matter:≤0.01 %
- ACS - Iron (Fe):≤0.002 %
- ACS - Loss on Drying at 105°C:≤0.2 %
- ACS - Nitrogen Compounds (as N):≤0.001 %
- ACS - pH of 5% Solution at 25°C:4.1 - 4.5
- ACS - Sodium (Na):≤0.005 %
- ACS - Sulfate (SO₄):≤0.003 %
- Solubility (10 in 100):Passes Test
- No Class 1,2,3 or other solvents are used or produced in the:
- manufacturing or purification of the product.:
- Elemental Impurities (USP 232, EP 5.20) - Information on elemental:
- impurities for this product is available on the associated Product:
- Regulatory Data Sheet and elemental impurity profile report.:
- J.T.Baker®, Potassium dihydrogen phosphate 98.0-100.5% (dried basis) NF, ACS:
Specification Test Results
About this item
Synonyms: Monopotassiumphosphate,Potassium phosphate monobasic,Monobasic Potassium Phosphate
:Length:
:Width:
:Height:
:Ø
:Thickness:
:98,0-100,5
:Management of Change (MOC) category = R
:Boiling Pt:
:Boiling Pt:
:Melting Pt:
:Melting Pt: 253 °C
:Flash Pt:
:Flash Pt:
:M.W. 136.09 g/mol
:253
:100
:GHS07
:12,07829
Formula:KH₂PO₄
MW:136.09 g/mol
MDL Number:MFCD00011401
CAS Number:7778-77-0
Customers Who Bought This Also Bought