Specifications
- Description:Fungi-Fluor® Kit for Fungal Detection
- Cat. no.:10819-090
- Supplier No.:17442
Specifications
About this item
The Fungi-Fluor® Kit for Fungal Detection offers a quick fluorescent stain/counterstain procedure for various fungal organisms
Kit contains 75mL of staining solution A, and 75mL of counterstaining solution B.
The kit can be used to screen a variety of specimen types, such as sputum and skin scrapings, for fungal detection.