Specifications
- Description:Fungi-Fluor® Pneumocystis Kit
- Cat. no.:10818-624
- No. of Reactions:100
- Supplier No.:22363
Specifications
About this item
The Fungi Fluor® Pneumocystis Kit offers a fast, fluorescent staining procedure for Pneumocystis jiroveci in bronchial specimens.
Kit contains 10mL of Fungi-Fluor® staining solution, 10 positive unstained control slides, and Direct fluorescent stain for Pneumocystis carinii cyst.