To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Control
- Antigen name:IgG2b, k Isotype Ctrl
- Clonality:Monoclonal
- Clone:MPC-11
- Host:Mouse
- Isotype:IgG2b kappa
- Reactivity:Mouse
- Antigen symbol:IgG2b, k Isotype Ctrl
- Conjugation:Brilliant Violet® 785
- Size:25 tests
- Form:Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide and BSA (origin USA).
- Storage temperature:The antibody solution should be stored undiluted between 2 °C and 8 °C, and protected from prolonged exposure to light. Do not freeze.
- Concentration:0.2 mg/ml
- Shipping temperature:Ambient RT
- Purification:The antibody was purified by affinity chromatography and conjugated with Brilliant Violet 785™ under optimal conditions. The solution is free of unconjugated Brilliant Violet 785™ and unconjugated antibody.
- Cat. no.:CA10761-132
- Supplier No.:400355
Specifications
About this item
Brilliant Violet 785™ Mouse IgG2b, κ Isotype Ctrl [MPC-11]; Isotype: Mouse IgG2b, κ; Reactivity: Mouse; Apps: FC, ICFC; Size: 100 tests
Each lot of this mouse IgG2b, κ isotype control antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, use the isotype control at the same concentration as your primary antibody. Use our Concentration Lookup tool to find the exact concentrations of your lots of product.
Type: Control
Antigen: IgG2b, k Isotype Ctrl
Clonality: Monoclonal
Clone: MPC-11
Conjugation: Brilliant Violet® 785
Epitope:
Host: Mouse
Isotype: Mouse IgG2b, κ
Reactivity: Mouse