Anti-DLG4 Mouse Monoclonal Antibody [clone: K28/74]
Catalog #: CA10816-652
Supplier: Biolegend
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:Discs, Large Homolog 4 (Drosophila)
- Clonality:Monoclonal
- Clone:K28/74
- Gene ID:1742
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human,Rat,Mouse
- Antigen symbol:DLG4
- ImmunoChemistry:Yes
- Size:100 µL
- Western blot:Yes
- Form:Phosphate-buffered solution.
- Antigen synonyms:Tax interaction protein 15|SAP-90|discs large homolog 4|SAP90|large homolog 4 (Drosophila)|FLJ98574|DLG4|synapse-associated protein 90|discs|PSD95|post-synaptic density protein 95|FLJ97752
- Storage temperature:Store at –20 °C. Upon initial thawing, apportion into working aliquots and store at –20 °C. Avoid repeated freeze-thaw cycles to prevent denaturing the antibody. For long-term storage, keep the antibody at –80 °C.
- Concentration:1 mg/ml
- Shipping temperature:Dry Ice
- Immunogen:This monoclonal antibody was raised against a fusion protein corresponding to amino acids 77-299 (PDZ domains 1 and 2) of human PSD-95.
- Purification:The antibody was purified by affinity chromatography.
- Cat. no.:CA10816-652
- Supplier No.:810301
Specifications
About this item
Type: Primary
Antigen: DLG4
Clonality: Monoclonal
Clone: K28/74
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Rat
Customers Who Bought This Also Bought