Anti-PPP3CA Mouse Monoclonal Antibody [clone: 2E2.D11]
Catalog #: CA10816-784
Supplier: Biolegend
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:Protein Phosphatase 3, Catalytic Subunit, Alpha Isozyme
- Clonality:Monoclonal
- Clone:2E2.D11
- Gene ID:5530
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human,Rat,Mouse
- Antigen symbol:PPP3CA
- ImmunoChemistry:Yes
- Size:100 µL
- Western blot:Yes
- Form:Phosphate-buffered solution.
- Antigen synonyms:CALN|CCN1|PPP2B|CNA1|CALNA1|CALNA
- Storage temperature:The antibody solution should be stored undiluted between 2°C and 8°C.
- Concentration:1 mg/ml
- Shipping temperature:Ambient RT
- Immunogen:This monoclonal antibody was raised against a peptide sequence conjugated to KLH corresponding to amino acids 478-492 of human CnA1.
- Purification:The antibody was purified by affinity chromatography.
- Cat. no.:CA10816-784
- Supplier No.:817601
Specifications
About this item
Type: Primary
Antigen: PPP3CA
Clonality: Monoclonal
Clone: 2E2.D11
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Rat
Customers Who Bought This Also Bought