Anti-LRRK2 Mouse Monoclonal Antibody [clone: MC.028.83.76.242]
Catalog #: CA10816-434
Supplier: Biolegend
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:Leucine-rich Repeat kinase 2
- Clonality:Monoclonal
- Clone:MC.028.83.76.242
- Gel shift:Yes
- Gene ID:120892
- Host:Mouse
- Isotype:IgG1
- Reactivity:Primates,Human,Dog,Rat,Mouse
- Antigen symbol:LRRK2
- ELISA:Yes
- ImmunoChemistry:Yes
- Size:100 µL
- Western blot:Yes
- Form:Phosphate-buffered solution.
- Antigen synonyms:Dardarin|LRRK 2|BC 300-268|PARK8|ROCO2|LRR kinase 2|Leucine-rich repeat kinase 2|LRRK-2
- Storage temperature:The antibody solution should be stored undiluted between 2°C and 8°C.
- Concentration:1 mg/ml
- Shipping temperature:Ambient RT
- Immunogen:This monoclonal antibody was raised against a peptide immunogen comprising amino acids 2069-2087 of the LRRK2 protein in mice that did not express endogenous LRRK2 protein.
- Purification:The antibody was purified by affinity chromatography.
- Cat. no.:CA10816-434
- Supplier No.:808201
Specifications
About this item
Type: Primary
Antigen: LRRK2
Clonality: Monoclonal
Clone: MC.028.83.76.242
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Rat, Dog, Primate
Customers Who Bought This Also Bought