To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:Plexin B2
- Clonality:Monoclonal
- Clone:3E7
- Gene ID:140570
- Host:Armenian hamster
- Isotype:IgG
- Reactivity:Mouse
- Antigen symbol:Plexin B2
- Conjugation:PE (Phycoerythrin)
- Size:50 µg
- Form:Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide.
- Antigen synonyms:PLEXB2|Debt|plexin-B2|PLXNB2|MM1
- Storage temperature:The antibody solution should be stored undiluted between 2 °C and 8 °C, and protected from prolonged exposure to light. Do not freeze.
- Concentration:0.20 mg/mL
- Shipping temperature:Ambient RT
- Immunogen:PAM 2-12 keratinocyets cell line
- Purification:The antibody was purified by affinity chromatography and conjugated with PE under optimal conditions. The solution is free of unconjugated PE and unconjugated antibody.
- Cat. no.:CA10756-504
- Supplier No.:145903
Specifications
About this item
PE anti-mouse Plexin B2 [3E7]; Isotype: Armenian Hamster IgG; Reactivity: Mouse; Apps: FC; Size: 50 μg
Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 0.25 μg per million cells in 100 μl volume. It is recommended that the reagent be titrated for optimal performance for each application.
Type: Primary
Antigen: Plexin B2
Clonality: Monoclonal
Clone: 3E7
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Armenian Hamster
Isotype: Armenian Hamster IgG
Reactivity: Mouse