To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Control
- Antigen name:IgY
- Clonality:Polyclonal
- Clone:Poly4021
- Gel shift:Yes
- Host:Chicken
- Isotype:IgY
- Reactivity:Mammals
- Antigen symbol:IgY
- ELISA:Yes
- ImmunoChemistry:Yes
- Size:25 µL
- Western blot:Yes
- Form:This isotype control is lyophilized.
- Antigen synonyms:NEU5GC
- Storage temperature:For long-term storage, store the lyophilized antibody at -20 °C or -70 °C. After reconstitution, store the antibody between 2 °C and 8 °C for up to 2 weeks, or store in aliquots at -20 °C for up to 6 months.
- Concentration:0.00 mg/mL
- Shipping temperature:Ambient RT
- Purification:Reconstitute the contents of the antibody vial by adding 25 µl of distilled water.
- Cat. no.:CA10761-252
- Supplier No.:402101
Specifications
About this item
Purified Chicken IgY Isotype Ctrl [Poly4021]; Isotype: Chicken Polyclonal IgY; Apps: FC; Size: 25 μl
Each lot of this isotype control antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, use the isotype control at the same concentration as your primary antibody. Use our Concentration Lookup tool to find the exact concentrations of your lots of product.
Type: Control
Antigen: IgY Isotype Ctrl
Clonality: Polyclonal
Clone: Poly4021
Conjugation:
Epitope:
Host: Chicken
Isotype: Chicken Polyclonal IgY
Reactivity: