To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:IL-21
- Clonality:Monoclonal
- Clone:3A3-N2
- Gel shift:Yes
- Gene ID:59067
- Host:Mouse
- Isotype:IgG1 kappa
- Reactivity:Rhesus monkey,Human
- Antigen symbol:IL-21
- ELISA:Yes
- Size:100 µg
- Form:Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide and EDTA.
- Antigen synonyms:IL21|Interleukin 21
- Storage temperature:The antibody solution should be stored undiluted between 2 °C and 8 °C.
- Concentration:1.00 mg/mL
- Shipping temperature:Ambient RT
- Immunogen:Recombinant full length human IL-21
- Purification:The antibody was purified by affinity chromatography.
- Cat. no.:CA10761-676
- Supplier No.:513009
Specifications
About this item
Purified anti-human IL-21 (MaxPar® Ready) [3A3-N2]; Isotype: Mouse IgG1, κ; Reactivity: Human, Cross-Reactivity: Rhesus; Apps: ELISA Capture, CyTOF®; Size: 100 μg
This product is suitable for use with the MaxPar® Metal Labeling Kits. The product is formulated so that the buffer exchange step is not required (steps 7, 8, 9, and 11 in the MaxPar® antibody labeling protocol). Just add 100 μl of this antibody to 100 μl of 4 mM TCEP-R in a 50 kDa filter, as described in step 12, and continue with the protocol.
Type: Primary
Antigen: IL-21
Clonality: Monoclonal
Clone: 3A3-N2
Conjugation:
Epitope:
Host: Mouse
Isotype: Mouse IgG1, κ
Reactivity: Human;Cross-Reactivity: Rhesus