About this item
PE/Dazzle™ 594 Mouse IgG2a, κ Isotype Ctrl [MOPC-173]; Isotype: Mouse IgG2a, κ; Reactivity: Mouse; Apps: FC, ICFC; Size: 100 μg
Each lot of this mouse IgG2a, κ isotype control antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, use the isotype control at the same concentration as your primary antibody. Use our Concentration Lookup tool to find the exact concentrations of your lots of product.
Type: Control
Antigen: IgG2a, k Isotype Ctrl
Clonality: Monoclonal
Clone: MOPC-173
Conjugation: PE (Phycoerythrin)/Dazzle 594
Epitope:
Host: Mouse
Isotype: Mouse IgG2a, κ
Reactivity: Mouse
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Catalog No:
- CA10761-114
- CA10761-116
- Antigen name:
- IgG2a, k Isotype Ctrl
- IgG2a, k Isotype Ctrl
- Antibody type:
- Control
- Control
- Clonality:
- Monoclonal
- Monoclonal
- Clone:
- MOPC-173
- MOPC-173
- Reactivity:
- Mouse
- Mouse
- Host:
- Mouse
- Mouse
- Isotype:
- IgG2a kappa
- IgG2a kappa