To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Control
- Antigen name:IgG2a, k Isotype Ctrl
- Clonality:Monoclonal
- Clone:RTK2758
- Host:Rat
- Isotype:IgG2a kappa
- Antigen symbol:IgG2a, k Isotype Ctrl
- Conjugation:Alexa Fluor® 594
- ImmunoFluorescence:Yes
- Size:100 µg
- Form:Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide.
- Storage temperature:The antibody solution should be stored undiluted between 2 °C and 8 °C, and protected from prolonged exposure to light. Do not freeze.
- Concentration:0.50 mg/mL
- Shipping temperature:Ambient RT
- Immunogen:Trinitrophenol + KLH
- Purification:The antibody was purified by affinity chromatography and conjugated with Alexa Fluor® 594 under optimal conditions. The solution is free of unconjugated Alexa Fluor® 594.
- Cat. no.:CA10761-178
- Supplier No.:400555
Specifications
About this item
Alexa Fluor® 594 Rat IgG2a, κ Isotype Ctrl [RTK2758]; Isotype: Rat IgG2a, κ; Reactivity: KLH; Apps: IF; Size: 100 μg
Each lot of this rat IgG2a, κ isotype control antibody is quality control tested by immunofluorescent staining. Use at concentrations comparable to those of the specific antibody of interest.
Type: Control
Antigen: IgG2a, k Isotype Ctrl
Clonality: Monoclonal
Clone: RTK2758
Conjugation: Alexa Fluor® 594
Epitope:
Host: Rat
Isotype: Rat IgG2a, κ
Reactivity: KLH