About this item
Brilliant Violet 650™ Rat IgG2a, κ isotype Ctrl [RTK2758]; Isotype: Rat IgG2a, κ; Reactivity: KLH; Apps: FC, ICFC; Size: 50 μg
Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis as negative control. Use at concentrations comparable to those of the specific antibody of interest.
Type: Control
Antigen: IgG2a, k Isotype Ctrl
Clonality: Monoclonal
Clone: RTK2758
Conjugation: Brilliant Violet® 650
Epitope:
Host: Rat
Isotype: Rat IgG2a, κ
Reactivity: KLH
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Catalog No:
- CA400541-BL
- CA10761-166
- Antigen symbol:
- IgG2a, k Isotype Ctrl
- IgG2a, k Isotype Ctrl
- Antigen name:
- IgG2a, k Isotype Ctrl
- IgG2a, k Isotype Ctrl
- Antibody type:
- Control
- Control
- Clonality:
- Monoclonal
- Monoclonal
- Clone:
- RTK2758
- RTK2758
- Host:
- Rat
- Rat
- Isotype:
- IgG2a kappa
- IgG2a kappa